COMH4211B808 Combi-Blocks, Inc.

Z-Ala-leu-nh2

  • MDL number MFCD00026502
  • Cas Number : 18323-56-3
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5022

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5022
Availability details for
Quantity
  • Availability
  • Add to Cart