COMH4211B80C Combi-Blocks, Inc.

4-Phenoxybutanal

  • MDL number MFCD16824615
  • Cas Number : 19790-62-6
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5037
Availability details for
Quantity
  • Availability
  • Add to Cart