COMH4211B80D Combi-Blocks, Inc.

3'-Methyl-4'-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-biphenyl-4-carboxylic acid methyl ester

  • MDL number MFCD29067530
  • Cas Number : 1411993-04-8
  • Molecular Weight : 352.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5043

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5043
Availability details for
Quantity
  • Availability
  • Add to Cart