COMH4211B80E Combi-Blocks, Inc.

2-[(2-Bromoethyl)sulfanyl]thiophene

  • MDL number MFCD11156121
  • Cas Number : 55697-88-6
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5044
Availability details for
Quantity
  • Availability
  • Add to Cart