COMH4211B80F Combi-Blocks, Inc.

2-Bromo-4-tert-butyl-5-nitro-phenylamine

  • MDL number MFCD20485042
  • Cas Number : 873055-28-8
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5046
Availability details for
Quantity
  • Availability
  • Add to Cart