COMH4211B810 Combi-Blocks, Inc.

Chloromethyl cyclohexanecarboxylate

  • MDL number MFCD20622050
  • Cas Number : 29916-48-1
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5048
Availability details for
Quantity
  • Availability
  • Add to Cart