COMH4211B811 Combi-Blocks, Inc.

2-(3-Bromo-5-chlorophenyl)propanoic acid

  • MDL number MFCD26658611
  • Cas Number : 1161361-93-8
  • Molecular Weight : 263.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5049
Availability details for
Quantity
  • Availability
  • Add to Cart