COMH4211B812 Combi-Blocks, Inc.

Dipyridamole impurity e

  • MDL number MFCD27965987
  • Cas Number : 2365420-11-5
  • Molecular Weight : 504.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5051
Availability details for
Quantity
  • Availability
  • Add to Cart