COMH4211B816 Combi-Blocks, Inc.

4-(4-Carboxy-3-trifluoromethyl-phenyl)-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583150
  • Cas Number : 2270912-87-1
  • Molecular Weight : 371.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5058
Availability details for
Quantity
  • Availability
  • Add to Cart