COMH4211B817 Combi-Blocks, Inc.

4-(2,2-Difluoro-propoxy)-3-fluoro-benzoic acid

  • MDL number MFCD31539544
  • Cas Number : 2157958-67-1
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5061
Availability details for
Quantity
  • Availability
  • Add to Cart