COMH4211B818 Combi-Blocks, Inc.

1-(2-Bromo-5-fluoro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561532
  • Cas Number : 1599089-24-3
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5063
Availability details for
Quantity
  • Availability
  • Add to Cart