COMH4211B81A Combi-Blocks, Inc.

4,4-Difluoro-1-[(3-iodophenyl)carbonyl]piperidine

  • MDL number MFCD31381586
  • Cas Number : 1228774-41-1
  • Molecular Weight : 351.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5065
Availability details for
Quantity
  • Availability
  • Add to Cart