COMH4211B81B Combi-Blocks, Inc.

(4-tert-Butoxycarbonylmethoxy-phenyl)-acetic acid

  • MDL number MFCD31618883
  • Cas Number : 119293-45-7
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5066
Availability details for
Quantity
  • Availability
  • Add to Cart