COMH4211B81D Combi-Blocks, Inc.

(5-Bromo-thiophen-3-yl)-piperidin-1-yl-methanone

  • MDL number MFCD12145321
  • Cas Number : 1147327-26-1
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5073
Availability details for
Quantity
  • Availability
  • Add to Cart