COMH4211B820 Combi-Blocks, Inc.

Benzene, 4-bromo-2-(3-buten-1-yloxy)-1-fluoro-

  • MDL number MFCD31381604
  • Cas Number : 1062615-67-1
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5078
Availability details for
Quantity
  • Availability
  • Add to Cart