COMH4211B821 Combi-Blocks, Inc.

Methyl 2-hydroxy-4-(2-methoxyethoxy)benzoate

  • MDL number MFCD20129536
  • Cas Number : 553672-08-5
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5079
Availability details for
Quantity
  • Availability
  • Add to Cart