COMH4211B825 Combi-Blocks, Inc.

Benzo[1,3]dioxol-4-ylmethyl-cyclopropyl-amine

  • MDL number MFCD12143039
  • Cas Number : 1049164-17-1
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5115
Availability details for
Quantity
  • Availability
  • Add to Cart