COMH4211B826 Combi-Blocks, Inc.

5-Chloro-4-fluoro-2-iodophenol

  • MDL number MFCD28972960
  • Cas Number : 1235407-13-2
  • Molecular Weight : 272.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5117
Availability details for
Quantity
  • Availability
  • Add to Cart