COMH4211B828 Combi-Blocks, Inc.

5-Amino-2,4-difluoro-n-methoxybenzamide

  • MDL number MFCD24624163
  • Cas Number : 658085-51-9
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5121
Availability details for
Quantity
  • Availability
  • Add to Cart