COMH4211B829 Combi-Blocks, Inc.

4-(4-Amino-2-methyl-phenoxy)-benzoic acid

  • MDL number MFCD31561588
  • Cas Number : 2270906-46-0
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5122
Availability details for
Quantity
  • Availability
  • Add to Cart