COMH4211B82A Combi-Blocks, Inc.

5-Fluoro-2-(1h-1,2,4-triazol-1-yl)benzonitrile

  • MDL number MFCD18860663
  • Cas Number : 449758-80-9
  • Molecular Weight : 188.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5124
Availability details for
Quantity
  • Availability
  • Add to Cart