COMH4211B82C Combi-Blocks, Inc.

2,2-Dimethyl-3-(3-methylphenoxy)propanoic acid

  • MDL number MFCD09704682
  • Cas Number : 938116-67-7
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-dimethyl-3-(3-methylphenoxy)propanoic acid
Inchi Code 1S/C12H16O3/c1-9-5-4-6-10(7-9)15-8-12(2,3)11(13)14/h4-7H,8H2,1-3H3,(H,13,14)
Inchi Key SUVJPLJPCOHDBK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5127
Availability details for
Quantity
  • Availability
  • Add to Cart