COMH4211B82D Combi-Blocks, Inc.

1-[(3-Fluorophenyl)acetyl]piperidine-3-carboxylic acid

  • MDL number MFCD09806570
  • Cas Number : 1016724-44-9
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5132
Availability details for
Quantity
  • Availability
  • Add to Cart