COMH4211B82E Combi-Blocks, Inc.

3-[(4-Hydroxy-pyrazole-1-carbonyl)-amino]-propionic acid methyl ester

  • MDL number MFCD31583144
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5133
Availability details for
Quantity
  • Availability
  • Add to Cart