COMH4211B830 Combi-Blocks, Inc.

5-Amino-n,n-diethyl-2-methylbenzamide

  • MDL number MFCD14618164
  • Cas Number : 251552-32-6
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5141
Availability details for
Quantity
  • Availability
  • Add to Cart