COMH4211B832 Combi-Blocks, Inc.

3-(3-Methyl-butyl)-benzoic acid

  • MDL number MFCD20641517
  • Cas Number : 105337-81-3
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5148
Availability details for
Quantity
  • Availability
  • Add to Cart