COMH4211B834 Combi-Blocks, Inc.

3-Amino-5-methoxy-n-methylbenzamide

  • MDL number MFCD23719970
  • Cas Number : 1346245-37-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5150
Availability details for
Quantity
  • Availability
  • Add to Cart