COMH4211B836 Combi-Blocks, Inc.

5-Chloro-2-(4-(trifluoromethoxy)benzyloxy)benzaldehyde

  • MDL number MFCD31557801
  • Cas Number : 1835283-96-9
  • Molecular Weight : 330.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5153
Availability details for
Quantity
  • Availability
  • Add to Cart