COMH4211B837 Combi-Blocks, Inc.

4-Bromo-n-cyclopropyl-3-fluorobenzenesulfonamide

  • MDL number MFCD28805504
  • Cas Number : 1055995-84-0
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5154
Availability details for
Quantity
  • Availability
  • Add to Cart