COMH4211B838 Combi-Blocks, Inc.

4-Bromo-n-methoxy-n,2-dimethylbenzamide

  • MDL number MFCD11847399
  • Cas Number : 178313-45-6
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5155

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5155
Availability details for
Quantity
  • Availability
  • Add to Cart