COMH4211B83A Combi-Blocks, Inc.

4,4-Difluoro-1-(4-methylsulfanyl-phenyl)-butane-1,3-dione

  • MDL number MFCD11188532
  • Cas Number : 134731-37-6
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5158
Availability details for
Quantity
  • Availability
  • Add to Cart