COMH4211B83B Combi-Blocks, Inc.

1-(4-Fluoro-3-methylbenzoyl)piperazine

  • MDL number MFCD18293678
  • Cas Number : 1267272-39-8
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5159
Availability details for
Quantity
  • Availability
  • Add to Cart