COMH4211B83C Combi-Blocks, Inc.

2-(Phenylmethoxy)benzoic acid 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD29079344
  • Cas Number : 287971-19-1
  • Molecular Weight : 325.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5163
Availability details for
Quantity
  • Availability
  • Add to Cart