COMH4211B83D Combi-Blocks, Inc.

(2-Ethoxy-3-fluorophenyl)methanamine

  • MDL number MFCD26055814
  • Cas Number : 1532017-59-6
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-ethoxy-3-fluorophenyl)methanamine
Inchi Code 1S/C9H12FNO/c1-2-12-9-7(6-11)4-3-5-8(9)10/h3-5H,2,6,11H2,1H3
Inchi Key NEKHTLOUVHNEHO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5164
Availability details for
Quantity
  • Availability
  • Add to Cart