COMH4211B83F Combi-Blocks, Inc.

2-Fluoro-4-(propan-2-yloxy)benzonitrile

  • MDL number MFCD23114350
  • Cas Number : 1369881-23-1
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5167
Availability details for
Quantity
  • Availability
  • Add to Cart