COMH4211B845 Combi-Blocks, Inc.

Methyl 4-isocyanatobicyclo[2.2.2]octane-1-carboxylate

  • MDL number MFCD28118871
  • Cas Number : 1357397-18-2
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5180
Availability details for
Quantity
  • Availability
  • Add to Cart