COMH4211B847 Combi-Blocks, Inc.

1-(Cyclohexylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD28718236
  • Cas Number : 1430750-95-0
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5183
Availability details for
Quantity
  • Availability
  • Add to Cart