COMH4211B849 Combi-Blocks, Inc.

tert-Butyl n-(2,2-dimethoxyethyl)sulfamoylcarbamate

  • MDL number MFCD31381668
  • Cas Number : 2168999-32-2
  • Molecular Weight : 284.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5189
Availability details for
Quantity
  • Availability
  • Add to Cart