COMH4211B84B Combi-Blocks, Inc.

1-(2-Bromo-5-nitro-benzyl)-4,4-difluoro-piperidine

  • MDL number MFCD31614012
  • Cas Number : 2270912-52-0
  • Molecular Weight : 335.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5195
Availability details for
Quantity
  • Availability
  • Add to Cart