COMH4211B84D Combi-Blocks, Inc.

5-Bromo-2-cyclopropylamino-nicotinic acid tert-butyl ester

  • MDL number MFCD31561473
  • Cas Number : 2270906-81-3
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5199
Availability details for
Quantity
  • Availability
  • Add to Cart