COMH4211B84E Combi-Blocks, Inc.

1-Bromo-4-(4,4,4-trifluorobutoxy)benzene

  • MDL number MFCD25218933
  • Cas Number : 622387-06-8
  • Molecular Weight : 283.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5200
Availability details for
Quantity
  • Availability
  • Add to Cart