COMH4211B850 Combi-Blocks, Inc.

1-(4-Aminophenyl)-3-ethylurea

  • MDL number MFCD11106238
  • Cas Number : 70826-92-5
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5206
Availability details for
Quantity
  • Availability
  • Add to Cart