COMH4211B851 Combi-Blocks, Inc.

6-Bromo-n-(propan-2-yl)pyridine-3-carboxamide

  • MDL number MFCD21781802
  • Cas Number : 1254123-86-8
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5209
Availability details for
Quantity
  • Availability
  • Add to Cart