COMH4211B852 Combi-Blocks, Inc.

(E/Z)-3-(2-(Trifluoromethyl)-4-fluorophenyl)-acrylonitrile

  • MDL number MFCD25643454
  • Cas Number : 412961-55-8
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5211
Availability details for
Quantity
  • Availability
  • Add to Cart