COMH4211B854 Combi-Blocks, Inc.

4-Hydroxy-n,2-dimethylbenzamide

  • MDL number MFCD16998317
  • Cas Number : 50639-10-6
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5214
Availability details for
Quantity
  • Availability
  • Add to Cart