COMH4211B856 Combi-Blocks, Inc.

5-(2-Chloro-4-iodophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD22564215
  • Cas Number : 1388028-93-0
  • Molecular Weight : 337.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2-chloro-4-iodophenyl)-1,3,4-thiadiazol-2-amine
Inchi Code 1S/C8H5ClIN3S/c9-6-3-4(10)1-2-5(6)7-12-13-8(11)14-7/h1-3H,(H2,11,13)
Inchi Key ZWJXXDASJKTTLM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5216
Availability details for
Quantity
  • Availability
  • Add to Cart