COMH4211B857 Combi-Blocks, Inc.

2-(2-Fluoro-5-methyl-phenoxymethyl)-oxirane

  • MDL number MFCD20429783
  • Cas Number : 1343663-06-8
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5223

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5223
Availability details for
Quantity
  • Availability
  • Add to Cart