COMH4211B859 Combi-Blocks, Inc.

3-[2-(2-Fluoro-4-methyl-phenyl)-ethyl]-benzoic acid

  • MDL number MFCD31614032
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5227
Availability details for
Quantity
  • Availability
  • Add to Cart