COMH4211B85C Combi-Blocks, Inc.

1-(2,4-Dimethylbenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08443240
  • Cas Number : 1102854-98-7
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5234

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5234
Availability details for
Quantity
  • Availability
  • Add to Cart