COMH4211B85E Combi-Blocks, Inc.

4-(Cyclopropylmethoxy)-3,5-difluorobenzoic acid

  • MDL number MFCD18285388
  • Cas Number : 1343436-42-9
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(cyclopropylmethoxy)-3,5-difluorobenzoic acid
Inchi Code 1S/C11H10F2O3/c12-8-3-7(11(14)15)4-9(13)10(8)16-5-6-1-2-6/h3-4,6H,1-2,5H2,(H,14,15)
Inchi Key PFVZZPBKPNWGMO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5239
Availability details for
Quantity
  • Availability
  • Add to Cart